Assay Details
Gene Symbol: FPGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002085.4 | XP_004002134.3 | 2 - 3 | 339 | 90 | |
XM_012173799.2 | XP_012029189.2 | 2 - 3 | 453 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
