Assay Details
Gene Symbol: HDGFL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957110.1 | XP_027812911.1 | 4 - 5 | 575 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HDGFL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957110.1 | XP_027812911.1 | 4 - 5 | 575 | 103 |
Set Membership: |
![]() ![]() ![]() ![]() |