Assay Details
Gene Symbol: CDC14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004085.3 | XP_004004134.1 | 3 - 4 | 684 | 97 | |
XM_012116349.2 | XP_011971739.2 | 3 - 4 | 685 | 97 | ||
XM_012116425.2 | XP_011971815.1 | 3 - 4 | 684 | 97 | ||
XM_027964217.1 | XP_027820018.1 | 3 - 4 | 328 | 97 | ||
XM_027964218.1 | XP_027820019.1 | 3 - 4 | 328 | 97 | ||
XM_027964219.1 | XP_027820020.1 | 3 - 4 | 328 | 97 | ||
XM_027964220.1 | XP_027820021.1 | 3 - 4 | 328 | 97 | ||
XM_027964221.1 | XP_027820022.1 | 3 - 4 | 328 | 97 | ||
XM_027964222.1 | XP_027820023.1 | 3 - 4 | 328 | 97 |
Gene Symbol: LOC114108754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964727.1 | XP_027820528.1 | 3 - 4 | 237 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
