Assay Details
Gene Symbol: MIR221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107961.1 | 1 | 47 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | divergent protein kinase domain 2B | DIPK2B | CXHXorf36 | -438.4kb | Sense | Chr. X - 45748914 - 45789765 on NCBI Build Oar_rambouillet_v1.0 |
Downstream | serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit beta-like | LOC101108374 | 405.7kb | Anti-sense | Chr. X - 46633951 - 46651220 on NCBI Build Oar_rambouillet_v1.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
