Assay Details
Gene Symbol: CYP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123000.1 | NP_001116472.1 | 4 - 5 | 630 | 85 | |
GenBank mRNA | AJ012153.1 | - | 4 - 5 | 662 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
