Assay Details
Gene Symbol: CTSV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004081.3 | XP_004004130.1 | 4 - 5 | 526 | 102 | |
XM_004004082.4 | XP_004004131.1 | 4 - 5 | 558 | 102 | ||
XM_012116286.3 | XP_011971676.1 | 4 - 5 | 1183 | 102 | ||
XM_012116293.2 | XP_011971683.1 | 4 - 5 | 520 | 102 | ||
GenBank mRNA | DQ152935.1 | - | 2 - 3 | 254 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
