Assay Details
Gene Symbol: MYOZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199222.1 | NP_001186151.1 | 5 - 6 | 634 | 81 | |
XM_012179352.3 | XP_012034742.1 | 6 - 7 | 803 | 81 | ||
GenBank mRNA | HM628579.1 | - | 5 - 6 | 634 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
