Assay Details
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009397.1 | NP_001009397.1 | 1 - 2 | 569 | 70 | |
GenBank mRNA | L22215.1 | - | 1 - 2 | 556 | 70 | |
X72088.1 | - | 569 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
