Assay Details
Gene Symbol: SCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009254.1 | NP_001009254.1 | 5 - 6 | 1078 | 71 | |
GenBank mRNA | AJ001048.1 | - | 5 - 6 | 1078 | 71 | |
DQ239647.1 | - | 1 - 2 | 233 | 71 | ||
FJ542814.1 | - | 2 - 3 | 435 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
