Assay Details
Gene Symbol: IL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009783.1 | NP_001009783.1 | 3 - 4 | 346 | 105 | |
GenBank mRNA | U35038.1 | - | 3 - 4 | 346 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
