Assay Details
Gene Symbol: CYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009804.1 | NP_001009804.1 | 8 - 9 | 1019 | 114 | |
GenBank mRNA | X53037.1 | - | 1019 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
