Assay Details
Gene Symbol: ARF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162547.1 | NP_001156019.1 | 2 - 3 | 148 | 91 | |
GenBank mRNA | FJ969399.1 | - | 2 - 3 | 148 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
