Assay Details
Gene Symbol: TIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166186.1 | NP_001159658.1 | 2 - 3 | 228 | 122 | |
GenBank mRNA | GQ221040.1 | - | 2 - 3 | 228 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
