Assay Details
Gene Symbol: IFI30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205150.1 | NP_001192079.1 | 5 - 6 | 618 | 85 | |
GenBank mRNA | HM017967.1 | - | 5 - 6 | 618 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
