Assay Details
Gene Symbol: TLR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135930.1 | NP_001129402.1 | 2 - 3 | 267 | 100 | |
GenBank mRNA | AM981302.1 | - | 2 - 3 | 267 | 100 | |
AY957615.1 | - | 1 - 2 | 60 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
