Assay Details
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166199.1 | NP_001159671.1 | 2 - 3 | 329 | 84 | |
XM_012107898.3 | XP_011963288.1 | 2 - 3 | 364 | 84 | ||
XM_012108258.3 | XP_011963648.1 | 2 - 3 | 335 | 84 | ||
GenBank mRNA | GQ221043.1 | - | 2 - 3 | 329 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
