Assay Details
Gene Symbol: PRDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166200.1 | NP_001159672.1 | 4 - 5 | 514 | 80 | |
XM_012177564.2 | XP_012032954.1 | 5 - 6 | 674 | 80 | ||
XM_027969176.1 | XP_027824977.1 | 5 - 6 | 675 | 80 | ||
XM_027969177.1 | XP_027824978.1 | 5 - 6 | 721 | 80 | ||
GenBank mRNA | GQ221042.1 | - | 4 - 5 | 514 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
