Assay Details
Gene Symbol: ADIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306110.1 | NP_001293039.1 | 6 - 7 | 1002 | 104 | |
XM_027975558.1 | XP_027831359.1 | 5 - 6 | 3538 | 104 | ||
XM_027975559.1 | XP_027831360.1 | 6 - 7 | 2919 | 104 | ||
XM_027975560.1 | XP_027831361.1 | 5 - 6 | 3470 | 104 | ||
GenBank mRNA | KJ159212.1 | - | 6 - 7 | 1002 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
