Assay Details
Gene Symbol: TSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009747.1 | NP_001009747.1 | 1 - 2 | 174 | 82 | |
XM_027965743.1 | XP_027821544.1 | 2 - 3 | 295 | 82 | ||
GenBank mRNA | AF290203.1 | - | 1 - 2 | 174 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
