Assay Details
Gene Symbol: IFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009803.1 | NP_001009803.1 | 2 - 3 | 233 | 92 | |
GenBank mRNA | DQ028767.1 | - | 2 - 3 | 92 | ||
DQ410715.1 | - | 2 - 3 | 181 | 92 | ||
X52640.1 | - | 233 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
