Assay Details
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280707.1 | NP_001267636.1 | 2 - 3 | 305 | 77 | |
XM_015102519.2 | XP_014958005.2 | 3 - 4 | 351 | 77 | ||
GenBank mRNA | GU585526.1 | - | 2 - 3 | 305 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
