Assay Details
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009194.1 | NP_001009194.1 | 3 - 4 | 641 | 69 | |
GenBank mRNA | AF009037.1 | - | 3 - 4 | 641 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
