Assay Details
Gene Symbol: TTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198882.1 | NP_001185811.1 | 4 - 5 | 685 | 137 | |
GenBank mRNA | HQ268507.1 | - | 4 - 5 | 685 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
