Assay Details
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161862.1 | NP_001155334.1 | 5 - 6 | 423 | 97 | |
GenBank mRNA | FJ943970.1 | - | 5 - 6 | 423 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
