Assay Details
Gene Symbol: CIPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130933.1 | NP_001124405.1 | 2 - 3 | 309 | 67 | |
XM_027971430.1 | XP_027827231.1 | 3 - 4 | 1927 | 67 | ||
XM_027971431.1 | XP_027827232.1 | 3 - 4 | 593 | 67 | ||
XM_027971432.1 | XP_027827233.1 | 3 - 4 | 459 | 67 | ||
GenBank mRNA | EF683161.1 | - | 2 - 3 | 309 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
