Assay Details
Gene Symbol: RABAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161869.1 | NP_001155341.1 | 3 - 4 | 372 | 68 | |
GenBank mRNA | FJ943977.1 | - | 3 - 4 | 372 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
