Assay Details
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177678.1 | NP_001171149.1 | 2 - 3 | 600 | 99 | |
GenBank mRNA | GU437213.1 | - | 2 - 3 | 600 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
