Assay Details
Gene Symbol: OAZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290359.1 | NP_001277288.1 | 2 - 3 | 335 | 85 | |
NM_001301418.1 | NP_001288347.1 | 2 - 3 | 367 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
