Assay Details
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145180.1 | NP_001138652.1 | 3 - 4 | 418 | 102 | |
GenBank mRNA | FJ546079.1 | - | 3 - 4 | 418 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
