Assay Details
Gene Symbol: RAB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161881.1 | NP_001155353.1 | 1 - 2 | 162 | 80 | |
XM_015091600.2 | XP_014947086.1 | 2 - 3 | 648 | 80 | ||
XM_015091601.2 | XP_014947087.1 | 2 - 3 | 589 | 80 | ||
GenBank mRNA | FJ943983.1 | - | 1 - 2 | 162 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
