Assay Details
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078660.1 | NP_001072128.1 | 2 - 3 | 150 | 89 | |
XM_027977374.1 | XP_027833175.1 | 3 - 4 | 296 | 89 | ||
XM_027977375.1 | XP_027833176.1 | 3 - 4 | 322 | 89 | ||
XM_027977376.1 | XP_027833177.1 | 3 - 4 | 296 | 89 | ||
GenBank mRNA | DQ496115.1 | - | 2 - 3 | 150 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
