Assay Details
Gene Symbol: QPCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166178.1 | NP_001159650.1 | 4 - 5 | 725 | 132 | |
GenBank mRNA | AF039441.1 | - | 3 - 4 | 511 | 132 | |
GQ221058.1 | - | 4 - 5 | 725 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
