Assay Details
Gene Symbol: MAP3K20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012133548.3 | XP_011988938.1 | 6 - 7 | 744 | 103 | |
XM_027964843.1 | XP_027820644.1 | 6 - 7 | 641 | 103 | ||
XM_027964844.1 | XP_027820645.1 | 6 - 7 | 526 | 103 | ||
XM_027964845.1 | XP_027820646.1 | 6 - 7 | 598 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
