Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021782.4 | XP_004021831.1 | 7 - 8 | 2252 | 95 | |
XM_012105930.3 | XP_011961320.1 | 7 - 8 | 2252 | 95 | ||
XM_012105933.3 | XP_011961323.1 | 7 - 8 | 2252 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
