Assay Details
Gene Symbol: CPEB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016877.4 | XP_004016926.1 | 1 - 2 | 2168 | 70 | |
XM_012096733.3 | XP_011952123.1 | 1 - 2 | 2168 | 70 | ||
XM_012096734.3 | XP_011952124.1 | 1 - 2 | 2168 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
