Assay Details
Gene Symbol: LOC101121602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012178613.3 | XP_012034003.2 | 21 - 22 | 3835 | 98 | |
XM_012178614.3 | XP_012034004.2 | 20 - 21 | 3741 | 98 | ||
XM_027970311.1 | XP_027826112.1 | 21 - 22 | 3835 | 98 | ||
XM_027970312.1 | XP_027826113.1 | 20 - 21 | 3778 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
