Assay Details
Gene Symbol: LOC101105780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101638.2 | XP_014957124.2 | 11 - 12 | 1917 | 64 | |
XM_027956516.1 | XP_027812317.1 | 10 - 11 | 2502 | 64 | ||
XM_027956517.1 | XP_027812318.1 | 10 - 11 | 2504 | 64 | ||
XM_027956518.1 | XP_027812319.1 | 11 - 12 | 1844 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
