Assay Details
Gene Symbol: LOC101105780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101638.2 | XP_014957124.2 | 12 - 13 | 2089 | 79 | |
XM_027956516.1 | XP_027812317.1 | 11 - 12 | 2674 | 79 | ||
XM_027956517.1 | XP_027812318.1 | 11 - 12 | 2676 | 79 | ||
XM_027956518.1 | XP_027812319.1 | 12 - 13 | 2016 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
