Assay Details
Gene Symbol: TSC22D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963356.1 | XP_027819157.1 | 2 - 3 | 584 | 75 | |
XM_027963357.1 | XP_027819158.1 | 2 - 3 | 617 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
