Assay Details
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002980.4 | XP_004003029.2 | 3 - 4 | 479 | 74 | |
XM_012095791.3 | XP_011951181.1 | 3 - 4 | 481 | 74 | ||
XM_027956447.1 | XP_027812248.1 | 3 - 4 | 406 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
