Assay Details
Gene Symbol: C1QTNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009997.4 | XP_004010046.1 | 2 - 3 | 636 | 100 | |
XM_004009998.4 | XP_004010047.1 | 2 - 3 | 537 | 100 | ||
XM_004009999.4 | XP_004010048.1 | 2 - 3 | 707 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
