Assay Details
Gene Symbol: LOC114118248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979000.1 | XP_027834801.1 | 2 - 3 | 483 | 136 |
Gene Symbol: SLC25A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092151.2 | XP_014947637.2 | 2 - 3 | 444 | 136 | |
XM_027972507.1 | XP_027828308.1 | 2 - 3 | 335 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
