Assay Details
Gene Symbol: VSIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022038.4 | XP_004022087.3 | 1 - 2 | 186 | 68 | |
XM_004022039.4 | XP_004022088.3 | 1 - 2 | 189 | 68 | ||
XM_012106592.3 | XP_011961982.2 | 1 - 2 | 188 | 68 | ||
XM_012106594.3 | XP_011961984.2 | 1 - 2 | 193 | 68 | ||
XM_012106596.3 | XP_011961986.2 | 1 - 2 | 191 | 68 | ||
XM_012106597.3 | XP_011961987.2 | 1 - 2 | 190 | 68 | ||
XM_027962852.1 | XP_027818653.1 | 1 - 2 | 194 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
