Assay Details
Gene Symbol: CRYBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972398.1 | XP_027828199.1 | 19 - 20 | 6006 | 75 | |
XM_027972399.1 | XP_027828200.1 | 18 - 19 | 5918 | 75 | ||
XM_027972400.1 | XP_027828201.1 | 18 - 19 | 5719 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
