Assay Details
Gene Symbol: LOC114112696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963845.1 | XP_027819646.1 | 4 - 5 | 639 | 85 |
Gene Symbol: LIPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959102.1 | XP_027814903.1 | 6 - 7 | 951 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
