Assay Details
Gene Symbol: RELL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008903.4 | XP_004008952.1 | 2 - 3 | 1030 | 83 | |
XM_027970343.1 | XP_027826144.1 | 2 - 3 | 1019 | 83 | ||
XM_027970344.1 | XP_027826145.1 | 2 - 3 | 1028 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
