Assay Details
Gene Symbol: LOC101118793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972290.1 | XP_027828091.1 | 11 - 12 | 1553 | 110 | |
XM_027972291.1 | XP_027828092.1 | 10 - 11 | 1532 | 110 | ||
XM_027972292.1 | XP_027828093.1 | 4 - 5 | 880 | 110 |
Gene Symbol: LOC114108752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972289.1 | XP_027828090.1 | 3 - 4 | 492 | 110 |
Gene Symbol: LOC114116052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972288.1 | XP_027828089.1 | 3 - 4 | 948 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
