Assay Details
Gene Symbol: ACRV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019520.4 | XP_004019569.1 | 2 - 3 | 593 | 59 | |
XM_027959717.1 | XP_027815518.1 | 3 - 4 | 401 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
