Assay Details
Gene Symbol: CDC14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004085.3 | XP_004004134.1 | 1 - 2 | 519 | 58 | |
XM_012116349.2 | XP_011971739.2 | 1 - 2 | 520 | 58 | ||
XM_012116425.2 | XP_011971815.1 | 1 - 2 | 519 | 58 |
Gene Symbol: LOC114108754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964727.1 | XP_027820528.1 | 1 - 2 | 72 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
