Assay Details
Gene Symbol: LOC101119648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011703.4 | XP_004011752.1 | 5 - 6 | 1387 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101119648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011703.4 | XP_004011752.1 | 5 - 6 | 1387 | 86 |
Set Membership: |
![]() ![]() ![]() ![]() |